2-Bromo-4-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A207719-10G A207719-5G A207719-250MG
103817-814EA 65 USD
103817-814 103848-498 76999-544 77086-784 77671-838
2-Bromo-4-(trifluoromethyl)pyrimidine 97%
2-Bromo-4-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₂BrF₃N₂
MW: 226.9820 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09802248
CAS Number: 785777-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR