2-Bromo-4-(trifluoromethyl)pyridine 98%
Supplier: Ambeed
|
Warning
|
A170449-1G
A170449-25G
A170449-5G
A170449-100G
A170449-10G
A170449-500G
103806-502EA
7
USD
103806-502
103844-776
77081-884
77081-886
77081-888
77151-258
77151-260
77151-262
2-Bromo-4-(trifluoromethyl)pyridine 98%
2-Bromo-4-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₃BrF₃N MW: 226 g/mol Boiling Pt: 195.9 °C (760 mm Hg) Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153085 CAS Number: 175205-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...