2-Bromo-4-(trifluoromethyl)benzaldehyde 95%

Supplier: Ambeed
Warning

A296687-1G A296687-25G A296687-5G A296687-100G A296687-10G
103814-110EA 6 USD
103814-110 103835-252 76837-928 76837-930 76837-932 77262-918 77262-920
2-Bromo-4-(trifluoromethyl)benzaldehyde 95%
2-Bromo-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
MW: 253.0200 g/mol
Boiling Pt: 230 °C (760 mm Hg)
Flash Pt: 92.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09835101
CAS Number: 85118-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR