2-Bromo-4-(tert-butyl)aniline 96%

Supplier: Ambeed
Warning

A417724-100G A417724-10G A417724-25G A417724-1G A417724-5G
103827-340EA 6 USD
103827-340 103838-386 76694-952 76694-954 76841-976 77307-190 77307-192
2-Bromo-4-(tert-butyl)aniline 96%
2-Bromo-4-(tert-butyl)aniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN
MW: 228.13 g/mol
Boiling Pt: 265.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00052693
CAS Number: 103273-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR