2-Bromo-4-(bromomethyl)-1-methylbenzene 95%

Supplier: Ambeed
Danger

A509136-250MG A509136-1G A509136-5G
103828-362EA 124 USD
103828-362 103839-462 76830-278 77307-644 77307-646
2-Bromo-4-(bromomethyl)-1-methylbenzene 95%
2-Bromo-4-(bromomethyl)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂
Storage Temperature: Refrigerator
MDL Number: MFCD12025350
CAS Number: 259231-26-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR