2-Bromo-4-(4-methylpiperazin-1-yl)aniline 97%

Supplier: Ambeed
Warning

A1144282-100MG A1144282-250MG
103798-402EA 267 USD
103798-402 76673-152 76673-154
2-Bromo-4-(4-methylpiperazin-1-yl)aniline 97%
2-Bromo-4-(4-methylpiperazin-1-yl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₆BrN₃
MW: 270.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25959849
CAS Number: 166818-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR