2-Bromo-3-nitrotoluene 98%

Supplier: Ambeed
Warning

A375959-25G A375959-5G A375959-10G A375959-500G A375959-1G
103826-576EA 9 USD
103826-576 103837-600 76840-902 76840-904 77142-532 77142-534 77306-872
2-Bromo-3-nitrotoluene 98%
2-Bromo-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
Boiling Pt: 282.4 °C (760 mm Hg)
Melting Pt: 38…41 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134555
CAS Number: 41085-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR