2-Bromo-3-nitropyridine 97%

Supplier: Ambeed
Warning

A794183-25G A794183-5G A794183-100G A794183-10G A794183-1G
103841-032EA 6 USD
103841-032 76845-274 76845-276 77293-750 77293-752 77308-344
2-Bromo-3-nitropyridine 97%
2-Bromo-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
Boiling Pt: 244 °C (760 mm Hg)
Melting Pt: 122…125 °C
Flash Pt: 101.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00955613
CAS Number: 19755-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR