2-Bromo-3-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A145359-10G A145359-250MG A145359-1G A145359-5G
103803-524EA 29 USD
103803-524 103841-610 76811-948 76811-950 77316-892 77316-894
2-Bromo-3-nitrobenzaldehyde 95%
2-Bromo-3-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD15527595
CAS Number: 90407-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR