2-Bromo-3-nitro-5-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A112308-100MG A112308-10G A112308-250MG A112308-1G
103797-996EA 77 USD
103797-996 103823-830 77025-822 77025-824 77025-826 77310-636
2-Bromo-3-nitro-5-(trifluoromethyl)pyridine 95%
2-Bromo-3-nitro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂BrF₃N₂O₂
MW: 270.99 g/mol
Boiling Pt: 243 °C (760 mm Hg)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14702306
CAS Number: 1214336-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR