2-Bromo-3-methylpyrazine 95%

Supplier: Ambeed
Warning

A347159-100MG A347159-10G A347159-250MG A347159-1G A347159-5G
103825-960EA 32 USD
103825-960 103836-946 76788-342 76788-344 76788-346 76840-020 76840-022
2-Bromo-3-methylpyrazine 95%
2-Bromo-3-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
MW: 173.0100 g/mol
Boiling Pt: 194.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08705769
CAS Number: 120984-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR