2-Bromo-m-cresol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-3-methylphenol
A289342-1G
A289342-5G
A289342-100G
A289342-100MG
A289342-10G
A289342-250MG
A289342-25G
103813-828EA
9
USD
103813-828
103834-968
76837-536
76837-538
77262-802
77262-804
77262-806
77262-808
77305-562
2-Bromo-m-cresol 98%
2-Bromo-m-cresol
Precaución: for Research Use Only
Formula:
C₇H₇BrO MW: 187.0300 g/mol Boiling Pt: 215.2 °C (760 mm Hg) Melting Pt: 58…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD09991797 CAS Number: 22061-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...