2-Bromo-m-toluonitrile 98%

Supplier: Ambeed
Warning

A567220-10G A567220-1G A567220-5G
103828-784EA 23 USD
103828-784 103839-926 76819-176 76844-046 77307-832
2-Bromo-m-toluonitrile 98%
2-Bromo-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MW: 196.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025664
CAS Number: 263159-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR