2-Bromo-m-toluic acid 97%

Supplier: Ambeed
Warning

A125553-100G A125553-10G A125553-25G A125553-500G A125553-5G
76688-372EA 71.09 USD
76688-372 76688-374 76688-376 76688-378 77298-060
2-Bromo-m-toluic acid 97%
2-Bromo-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 135…138 °C
Flash Pt: 140.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00079721
CAS Number: 53663-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR