2-Bromo-3-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A226784-1G A226784-25G A226784-5G A226784-100MG A226784-10G A226784-250MG A226784-100G
103819-270EA 20 USD
103819-270 103850-178 77088-930 77088-932 77088-934 77165-406 77165-408 77165-410 77892-367
2-Bromo-3-methylbenzaldehyde 98%
2-Bromo-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Storage Temperature: Refrigerator
MDL Number: MFCD18392002
CAS Number: 109179-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR