2-Bromo-m-toluidine 98%

Supplier: Ambeed
Warning

A209123-100G A209123-10G A209123-25G A209123-5G A209123-1G
103817-918EA 6 USD
103817-918 103848-604 76785-632 76785-634 77086-944 77086-946 77303-086
2-Bromo-m-toluidine 98%
2-Bromo-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 244.7 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD06411367
CAS Number: 54879-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR