2-Bromo-3-methoxypyridine 98%

Supplier: Ambeed
Warning

A293039-1G A293039-10G A293039-25G A293039-100G
103813-966EA 9 USD
103813-966 103835-106 76837-718 77114-302 77305-622 77685-834
2-Bromo-3-methoxypyridine 98%
2-Bromo-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 233.4 °C (760 mm Hg)
Melting Pt: 45…49 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01570896
CAS Number: 24100-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR