2-Bromo-3-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A115448-10G A115448-250MG A115448-25G A115448-1G
103798-566EA 15 USD
103798-566 103824-422 77257-910 77257-912 77257-914 77311-242
2-Bromo-3-methoxybenzoic acid 98%
2-Bromo-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
Flash Pt: 153.8 °C
Storage Temperature: Ambient
MDL Number: MFCD11848447
CAS Number: 88377-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR