2-Bromo-3-iodopyridine 98%

Supplier: Ambeed
Warning

A293621-10G A293621-250MG A293621-1G A293621-5G A293621-25G A293621-100G
103813-988EA 12 USD
103813-988 103835-128 76787-596 76787-598 76787-600 76837-748 76837-750 77601-308 77872-379
2-Bromo-3-iodopyridine 98%
2-Bromo-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃BrIN
Boiling Pt: 284.1 °C (760 mm Hg)
Melting Pt: 95…100 °C
Flash Pt: 125.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09056782
CAS Number: 265981-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR