2-Bromo-3-hydroxypyridine 98%

Supplier: Ambeed
Warning

A123858-100G A123858-10G A123858-1G A123858-25G A123858-500G A123858-5G
76983-356EA 61 USD
76983-356 76983-358 76983-360 76983-362 76983-364 77312-576
2-Bromo-3-hydroxypyridine 98%
2-Bromo-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄BrNO
Boiling Pt: 321.7 °C (760 mm Hg)
Melting Pt: 185…188 °C
Flash Pt: 148.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR