2-Bromo-3-hexylthiophene 98%

Supplier: Ambeed
Warning

A165359-10G A165359-25G A165359-5G A165359-1G
103805-958EA 6 USD
103805-958 103844-214 77259-766 77300-774 77300-776 77320-416
2-Bromo-3-hexylthiophene 98%
2-Bromo-3-hexylthiophene

Precaución: for Research Use Only

Formula: C₁₀H₁₅BrS
MW: 247.20 g/mol
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD09907959
CAS Number: 69249-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR