2-Bromo-3-fluorophenol 98%

Supplier: Ambeed
Warning

A131747-100G A131747-10G A131747-1G A131747-25G A131747-5G
103801-624EA 5 USD
103801-624 77094-996 77094-998 77314-136 77314-138 77314-140
2-Bromo-3-fluorophenol 98%
2-Bromo-3-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrFO
MW: 190.9978 g/mol
Boiling Pt: 196 °C (760 mm Hg)
Flash Pt: 72.424 °C
Storage Temperature: Ambient
MDL Number: MFCD04037220
CAS Number: 443-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR