1-Bromo-2-fluoro-6-nitrobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-fluoronitrobenzene, 2-Bromo-1-fluoro-3-nitrobenzene

A108260-100G A108260-250MG A108260-25G A108260-1G A108260-5G
103797-316EA 13 USD
103797-316 103823-096 77283-702 77283-704 77283-706 77309-938 77309-940
1-Bromo-2-fluoro-6-nitrobenzene 95%
1-Bromo-2-fluoro-6-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Boiling Pt: 222.3±20.0 °C (760 mm Hg)
Flash Pt: 88.3 °C
Storage Temperature: Ambient
MDL Number: MFCD07782080
CAS Number: 59255-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR