2-Bromo-3-fluoroaniline 98%

Supplier: Ambeed
Warning

A280978-1G A280978-25G A280978-100G A280978-10G A280978-250MG
103813-528EA 10 USD
103813-528 103853-480 76837-172 76837-174 77289-664 77289-666 77289-668
2-Bromo-3-fluoroaniline 98%
2-Bromo-3-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrFN
Melting Pt: 30…34 °C
Flash Pt: 136.2 °C
Storage Temperature: Ambient
MDL Number: MFCD07369915
CAS Number: 111721-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR