2-Bromo-3-dodecylthiophene 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-laurylthiophene

A164263-100G A164263-10G A164263-1G A164263-25G A164263-5G
103805-834EA 7 USD
103805-834 103844-094 77028-730 77028-732 77320-248 77320-250 77320-252
2-Bromo-3-dodecylthiophene 97%
2-Bromo-3-dodecylthiophene

Precaución: for Research Use Only

Formula: C₁₆H₂₇BrS
MW: 331.36 g/mol
Boiling Pt: 370.5 °C (760 mm Hg)
Flash Pt: 177.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10000891
CAS Number: 139100-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR