2-Bromo-3-chlorothiophene 95%

Supplier: Ambeed
Warning

A102299-100MG A102299-250MG A102299-1G A102299-5G
103796-356EA 85 USD
103796-356 103822-048 77257-282 77257-284 77308-932 77308-934
2-Bromo-3-chlorothiophene 95%
2-Bromo-3-chlorothiophene

Precaución: for Research Use Only

Formula: C₄H₂BrClS
Melting Pt: 43…48 °C
Flash Pt: 76.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043882
CAS Number: 77893-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR