2-Bromo-3-chloropyridine 97%

Supplier: Ambeed
Warning

A392051-25G A392051-5G A392051-100G A392051-10G A392051-500G
103837-932EA 5 USD
103837-932 76841-330 76841-332 76990-146 76990-148 76990-150
2-Bromo-3-chloropyridine 97%
2-Bromo-3-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClN
Melting Pt: 58…63 °C
Storage Temperature: Ambient
MDL Number: MFCD04039344
CAS Number: 96424-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR