2-Bromo-3-chlorobenzaldehyde 98%

Supplier: Ambeed
Warning

A170345-100MG A170345-10G A170345-250MG A170345-1G A170345-5G A170345-25G
103806-474EA 15 USD
103806-474 103844-750 77029-006 77029-008 77029-010 77081-862 77081-864 77301-042
2-Bromo-3-chlorobenzaldehyde 98%
2-Bromo-3-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO
Boiling Pt: 269 °C (760 mm Hg)
Flash Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10699536
CAS Number: 74073-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR