2-Bromo-3,5-difluoropyridine 98%

Supplier: Ambeed
Warning

A251754-1G A251754-25G A251754-5G A251754-10G
103812-008EA 14 USD
103812-008 103851-848 77091-240 77091-242 77091-244 77113-466
2-Bromo-3,5-difluoropyridine 98%
2-Bromo-3,5-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrF₂N
Storage Temperature: Refrigerator
MDL Number: MFCD03840197
CAS Number: 660425-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR