2-Bromo-3,5-difluoroaniline 95%

Supplier: Ambeed
Warning

A293123-1G A293123-5G A293123-100G A293123-10G A293123-25G
103813-968EA 9 USD
103813-968 103835-108 76837-720 76837-722 77099-370 77099-372 77099-374
2-Bromo-3,5-difluoroaniline 95%
2-Bromo-3,5-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrF₂N
MW: 208.003 g/mol
Boiling Pt: 223.7 °C (760 mm Hg)
Flash Pt: 89.1 °C
Storage Temperature: Ambient
MDL Number: MFCD16661040
CAS Number: 500357-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR