2-Bromo-3,4-dimethylpyridine 98%

Supplier: Ambeed
Warning

A168119-100MG A168119-250MG
103806-254EA 195 USD
103806-254 103844-512 76985-308 76985-310
2-Bromo-3,4-dimethylpyridine 98%
2-Bromo-3,4-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD06637418
CAS Number: 33204-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR