2-Bromo-3,4-difluorobenzoic acid 98%

Supplier: Ambeed
Warning

A199885-100MG A199885-10G A199885-250MG A199885-1G A199885-25G A199885-5G
103817-146EA 11 USD
103817-146 103847-772 77030-092 77030-094 77030-096 77085-826 77085-828 77085-830
2-Bromo-3,4-difluorobenzoic acid 98%
2-Bromo-3,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
MW: 236.998 g/mol
Boiling Pt: 292.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09835210
CAS Number: 170108-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR