2-Bromo-3-(trifluoromethyl)pyridine 98%
Supplier: Ambeed
|
Danger
|
A220362-25G
A220362-5G
A220362-10G
A220362-1G
103818-854EA
5
USD
103818-854
103849-736
77088-324
77088-326
77275-464
77303-552
2-Bromo-3-(trifluoromethyl)pyridine 98%
2-Bromo-3-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₃BrF₃N MW: 226 g/mol Boiling Pt: 194.3 °C (760 mm Hg) Melting Pt: 37…41 °C Flash Pt: 28 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00153084 CAS Number: 175205-82-0 UN: 2926 ADR: 4.1,6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...