2-Bromo-2,3-dihydro-1H-indene 95%

Supplier: Ambeed
Warning

A127773-100MG A127773-250MG A127773-1G A127773-5G
103801-008EA 56 USD
103801-008 76971-936 76971-938 77298-232 77313-274
2-Bromo-2,3-dihydro-1H-indene 95%
2-Bromo-2,3-dihydro-1H-indene

Precaución: for Research Use Only

Formula: C₉H₉Br
MW: 197.07 g/mol
Boiling Pt: 249.2±29.0 °C (760 mm Hg)
Flash Pt: 91.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784485
CAS Number: 17623-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR