2-Bromo-1-phenylethan-1-ol 95%

Supplier: Ambeed
Warning

A823422-10G A823422-250MG A823422-1G A823422-5G
103829-978EA 18 USD
103829-978 103841-144 76653-872 76653-874 76845-382 77308-402
2-Bromo-1-phenylethan-1-ol 95%
2-Bromo-1-phenylethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 261.6 °C (760 mm Hg)
Flash Pt: 135.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00540454
CAS Number: 2425-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR