2-Bromo-1-phenylethan-1-ol 98%

Supplier: Ambeed
Warning

A291061-1G A291061-10G A291061-250MG A291061-25G A291061-5G
103813-886EA 16 USD
103813-886 103835-028 76837-616 77099-330 77099-332 77099-334 77305-590
2-Bromo-1-phenylethan-1-ol 98%
2-Bromo-1-phenylethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 261.6 °C (760 mm Hg)
Flash Pt: 135.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00540454
CAS Number: 2425-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR