2-Bromo-1-naphthaldehyde 95%

Supplier: Ambeed
Warning

A172073-100MG A172073-250MG
103806-690EA 255 USD
103806-690 103844-976 76974-062 76974-064
2-Bromo-1-naphthaldehyde 95%
2-Bromo-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₇BrO
MW: 235.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17012542
CAS Number: 269066-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR