2-Bromo-4-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-methoxy-4-nitrobenzene

A121082-100G A121082-10G A121082-25G A121082-5G
76766-482EA 109.28 USD
76766-482 76766-484 76766-486 77312-142
2-Bromo-4-nitroanisole 98%
2-Bromo-4-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Boiling Pt: 306.3 °C (760 mm Hg)
Melting Pt: 104…106 °C
Flash Pt: 139.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00041242
CAS Number: 5197-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR