2-Bromo-3-nitroanisole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-1-methoxy-3-nitrobenzene
A275985-25G
A275985-5G
A275985-100G
A275985-10G
103813-286EA
7
USD
103813-286
103853-214
77093-068
77093-070
77141-058
77141-060
2-Bromo-3-nitroanisole 98%
2-Bromo-3-nitroanisole
Precaución: for Research Use Only
Formula:
C₇H₆BrNO₃ Boiling Pt: 273.2 °C (760 mm Hg) Melting Pt: 90…94 °C Flash Pt: 119.072 °C Storage Temperature: Ambient |
MDL Number:
MFCD07779272 CAS Number: 67853-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...