3-Bromo-4-iodotoluene 98%

Supplier: Ambeed
Warning

A135912-100G A135912-10G A135912-25G A135912-5G
76767-488EA 72 USD
76767-488 76767-490 77315-118 77315-120
3-Bromo-4-iodotoluene 98%
3-Bromo-4-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrI
Boiling Pt: 266.7 °C (760 mm Hg)
Flash Pt: 115.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00079718
CAS Number: 71838-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR