3-Bromo-4-iodoanisole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-iodo-4-methoxybenzene

A235325-10G A235325-250MG A235325-25G A235325-1G A235325-5G
103811-004EA 39 USD
103811-004 103850-770 76815-274 76815-276 76815-278 77089-778 77089-780
3-Bromo-4-iodoanisole 97%
3-Bromo-4-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
MW: 312.9304 g/mol
Boiling Pt: 284.9 °C (760 mm Hg)
Flash Pt: 126.1 °C
Storage Temperature: Ambient
MDL Number: MFCD12547848
CAS Number: 466639-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR