3-Bromo-4-iodobenzotrifluoride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-1-iodo-4-(trifluoromethyl)benzene
A342763-25G
A342763-10G
A342763-1G
103825-872EA
10
USD
103825-872
103836-846
76839-898
77106-678
77306-550
3-Bromo-4-iodobenzotrifluoride 97%
3-Bromo-4-iodobenzotrifluoride
Precaución: for Research Use Only
Formula:
C₇H₃BrF₃I MW: 350.9024 g/mol Melting Pt: 36…38.5 °C Flash Pt: 97.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08543938 CAS Number: 481075-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...