2-Bromo-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A164782-100MG A164782-250MG
103805-902EA 147 USD
103805-902 103844-160 76973-738 76973-740
2-Bromo-1H-pyrrolo[2,3-b]pyridine 95%
2-Bromo-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11559018
CAS Number: 1083181-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR