2-Bromoimidazole 98%

Supplier: Ambeed
Warning

A126080-100G A126080-10G A126080-250MG A126080-1G A126080-25G A126080-5G
103800-760EA 9 USD
103800-760 77108-858 77108-860 77108-862 77108-864 77312-936 77312-938 77312-940
2-Bromoimidazole 98%
2-Bromoimidazole

Precaución: for Research Use Only

Formula: C₃H₃BrN₂
Boiling Pt: 273.1 °C (760 mm Hg)
Melting Pt: 197…202 °C
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179526
CAS Number: 16681-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR