1-Bromo-2-fluoro-5-iodobenzene 97%

Supplier: Ambeed
Warning

A231608-100G A231608-10G A231608-1G A231608-25G A231608-5G
103810-784EA 5 USD
103810-784 103850-536 76786-282 76786-284 77089-426 77089-428 77089-430
1-Bromo-2-fluoro-5-iodobenzene 97%
1-Bromo-2-fluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
MW: 300.9 g/mol
Boiling Pt: 243.4 °C (760 mm Hg)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04116322
CAS Number: 811842-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR