3-Bromo-α,α,α,4-tetrafluorotoluene 98%

Supplier: Ambeed
Warning

A100698-100G A100698-500G A100698-25G
103807-762EA 205 USD
103807-762 103821-280 76765-128 76765-130 77595-696
3-Bromo-α,α,α,4-tetrafluorotoluene 98%
3-Bromo-α,α,α,4-tetrafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
Boiling Pt: 152.7 °C (760 mm Hg)
Flash Pt: 71.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00040938
CAS Number: 68322-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR