2-Bromo-3-fluoroanisole 98%

Supplier: Ambeed
Warning

A290412-1G A290412-5G A290412-10G A290412-25G
103813-866EA 9 USD
103813-866 103835-008 76837-588 76837-590 77114-256 77305-580
2-Bromo-3-fluoroanisole 98%
2-Bromo-3-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFO
Flash Pt: 68.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03788540
CAS Number: 446-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR