1-Bromo-2-chloro-5-fluorobenzene 98%

Supplier: Ambeed
Warning

A139223-100G A139223-10G A139223-25G A139223-5G
77095-376EA 36 USD
77095-376 77095-378 77315-794 77315-796
1-Bromo-2-chloro-5-fluorobenzene 98%
1-Bromo-2-chloro-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClF
Boiling Pt: 417.7 °C (760 mm Hg)
Melting Pt: 246 °C
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00061120
CAS Number: 201849-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR