2-Bromo-1-chloro-3-nitrobenzene 98%

Supplier: Ambeed
Warning

A135451-100G A135451-10G A135451-250MG A135451-1G A135451-25G A135451-5G
103802-158EA 6 USD
103802-158 76782-622 76782-624 76782-626 77314-984 77314-986 77314-988
2-Bromo-1-chloro-3-nitrobenzene 98%
2-Bromo-1-chloro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MW: 236.45 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD15527221
CAS Number: 19128-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR