2-Bromo-3-chlorotoluene 97%

Supplier: Ambeed
Warning

A171945-1G A171945-100G A171945-10G A171945-25G A171945-5G
103806-680EA 8 USD
103806-680 103844-962 77082-136 77138-436 77138-438 77301-172 77301-174
2-Bromo-3-chlorotoluene 97%
2-Bromo-3-chlorotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrCl
Boiling Pt: 229.1 °C (760 mm Hg)
Flash Pt: 104.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00060645
CAS Number: 69190-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR